The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006073.3 | NP_006064.2 | 38 - 39 | 2284 | 162 |
GenBank mRNA | CR749591.1 | - | 27 - 28 | 993 | 162 |
U18985.1 | - | 38 - 39 | 1988 | 162 |