The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004686.4 | NP_004677.3 | 11 - 12 | 1592 | 105 |
XM_006716414.3 | XP_006716477.2 | 6 - 7 | 1958 | 105 | |
XM_011544692.2 | XP_011542994.1 | 8 - 9 | 1031 | 105 | |
XM_011544693.2 | XP_011542995.1 | 7 - 8 | 1493 | 105 | |
XM_017013956.1 | XP_016869445.1 | 10 - 11 | 1281 | 105 | |
GenBank mRNA | AF073482.1 | - | 7 - 8 | 861 | 105 |
AK294468.1 | - | 11 - 12 | 1353 | 105 | |
AK303762.1 | - | 3 - 4 | 415 | 105 | |
AK303803.1 | - | 2 - 3 | 365 | 105 | |
AK316022.1 | - | 11 - 12 | 1402 | 105 | |
AK316283.1 | - | 11 - 12 | 1412 | 105 | |
BC130577.1 | - | 11 - 12 | 1378 | 105 | |
BC130579.1 | - | 11 - 12 | 1378 | 105 | |
CR749240.1 | - | 11 - 12 | 1390 | 105 |