Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR7
Entrez Gene ID
9108
Gene Name
myotubularin related protein 7
Gene Aliases
-
Chromosome Location
Chr.8: 17296794 - 17413531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004686.4 NP_004677.3 12 - 13 1733 82
XM_006716414.3 XP_006716477.2 7 - 8 2099 82
XM_011544692.2 XP_011542994.1 9 - 10 1172 82
XM_011544693.2 XP_011542995.1 8 - 9 1634 82
XM_017013956.1 XP_016869445.1 11 - 12 1422 82
GenBank mRNA AK294468.1 - 12 - 13 1494 82
AK303762.1 - 4 - 5 556 82
AK303803.1 - 3 - 4 506 82
AK316022.1 - 12 - 13 1543 82
AK316283.1 - 12 - 13 1553 82
BC130577.1 - 12 - 13 1519 82
BC130579.1 - 12 - 13 1519 82
CR749240.1 - 12 - 13 1531 82

View Full Product Details