Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR7
Entrez Gene ID
9108
Gene Name
myotubularin related protein 7
Gene Aliases
-
Chromosome Location
Chr.8: 17296794 - 17413531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004686.4 NP_004677.3 9 - 10 1339 106
XM_006716414.3 XP_006716477.2 4 - 5 1705 106
XM_011544692.2 XP_011542994.1 6 - 7 778 106
XM_011544693.2 XP_011542995.1 5 - 6 1240 106
XM_017013956.1 XP_016869445.1 8 - 9 1028 106
GenBank mRNA AF073482.1 - 5 - 6 608 106
AK294468.1 - 9 - 10 1100 106
AK303762.1 - 1 - 2 162 106
AK316022.1 - 9 - 10 1149 106
AK316283.1 - 9 - 10 1159 106
BC130577.1 - 9 - 10 1125 106
BC130579.1 - 9 - 10 1125 106
CR749240.1 - 9 - 10 1137 106

View Full Product Details