Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASET2
Entrez Gene ID
8635
Gene Name
ribonuclease T2
Gene Aliases
RNASE6PL, bA514O12.3
Chromosome Location
Chr.6: 166929509 - 166956589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK001769.1 - 3 - 4 726 94

View Full Product Details