Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKKS
Entrez Gene ID
8195
Gene Name
McKusick-Kaufman syndrome
Gene Aliases
BBS6, HMCS, KMS, MKS
Chromosome Location
Chr.20: 10404780 - 10434239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018848.3 NP_061336.1 3 - 4 1871 89
NM_170784.2 NP_740754.1 3 - 4 1761 89
GenBank mRNA AF221992.1 - 3 - 4 1716 89
AF221993.1 - 3 - 4 1868 89
AK025741.1 - 3 - 4 1696 89
AK291925.1 - 3 - 4 1710 89
AL157427.1 - 3 - 4 1701 89
BC028973.2 - 3 - 4 1664 89

View Full Product Details