The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006929.4 | NP_008860.4 | 18 - 19 | 2589 | 84 |
XM_011514815.2 | XP_011513117.1 | 18 - 19 | 2296 | 84 | |
GenBank mRNA | AB209605.1 | - | 15 - 16 | 2692 | 84 |
AK297230.1 | - | 16 - 17 | 1961 | 84 | |
AK299967.1 | - | 9 - 10 | 1120 | 84 | |
AK303448.1 | - | 14 - 15 | 1804 | 84 | |
BC015758.1 | - | 18 - 19 | 2202 | 84 | |
U09877.1 | - | 18 - 19 | 2311 | 84 | |
Z48796.1 | - | 18 - 19 | 2252 | 84 |