The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006929.4 | NP_008860.4 | 20 - 21 | 2873 | 58 |
XM_011514815.2 | XP_011513117.1 | 20 - 21 | 2580 | 58 | |
GenBank mRNA | AB209605.1 | - | 17 - 18 | 2976 | 58 |
AK297230.1 | - | 18 - 19 | 2245 | 58 | |
AK299967.1 | - | 11 - 12 | 1404 | 58 | |
AK303448.1 | - | 16 - 17 | 2088 | 58 | |
BC015758.1 | - | 20 - 21 | 2486 | 58 | |
BE395851.1 | - | 1 - 2 | 104 | 58 | |
U09877.1 | - | 20 - 21 | 2595 | 58 | |
Z48796.1 | - | 20 - 21 | 2536 | 58 |