The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006929.4 | NP_008860.4 | 4 - 5 | 745 | 85 |
XM_011514815.2 | XP_011513117.1 | 4 - 5 | 452 | 85 | |
GenBank mRNA | AB209605.1 | - | 4 - 5 | 401 | 85 |
AK293881.1 | - | 4 - 5 | 408 | 85 | |
AK303302.1 | - | 4 - 5 | 392 | 85 | |
BC015758.1 | - | 4 - 5 | 358 | 85 | |
U09877.1 | - | 4 - 5 | 467 | 85 | |
Z48796.1 | - | 4 - 5 | 408 | 85 |