Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKIV2L
Entrez Gene ID
6499
Gene Name
Ski2 like RNA helicase
Gene Aliases
170A, DDX13, HLP, SKI2, SKI2W, SKIV2, SKIV2L1, THES2
Chromosome Location
Chr.6: 31958804 - 31969852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006929.4 NP_008860.4 5 - 6 856 86
XM_011514815.2 XP_011513117.1 5 - 6 563 86
GenBank mRNA AB209605.1 - 5 - 6 512 86
AK293881.1 - 5 - 6 519 86
AK303302.1 - 5 - 6 503 86
BC015758.1 - 5 - 6 469 86
U09877.1 - 5 - 6 578 86
Z48796.1 - 5 - 6 519 86

View Full Product Details