The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205207.1 | NP_001192136.1 | 4 - 5 | 1195 | 106 |
NM_194255.2 | NP_919231.1 | 5 - 6 | 1446 | 106 | |
XM_005261164.3 | XP_005261221.1 | 5 - 6 | 1419 | 106 | |
XM_011529696.2 | XP_011527998.1 | 6 - 7 | 1878 | 106 | |
XM_011529698.2 | XP_011528000.1 | 5 - 6 | 1482 | 106 | |
XM_011529700.2 | XP_011528002.1 | 5 - 6 | 1741 | 106 | |
XM_011529701.2 | XP_011528003.1 | 6 - 7 | 1691 | 106 | |
XM_011529702.2 | XP_011528004.1 | 5 - 6 | 1405 | 106 | |
XM_011529703.2 | XP_011528005.1 | 5 - 6 | 1473 | 106 | |
XM_011529704.2 | XP_011528006.1 | 5 - 6 | 1389 | 106 | |
XM_011529706.2 | XP_011528008.1 | 4 - 5 | 1834 | 106 | |
XM_011529709.2 | XP_011528011.1 | 5 - 6 | 1660 | 106 | |
XM_017028443.1 | XP_016883932.1 | 5 - 6 | 1570 | 106 | |
GenBank mRNA | AF004354.1 | - | 4 - 5 | 1364 | 106 |
AK303168.1 | - | 4 - 5 | 1182 | 106 | |
AK307623.1 | - | 5 - 6 | 1450 | 106 | |
AK313125.1 | - | 5 - 6 | 1410 | 106 | |
S78996.1 | - | 4 - 5 | 1293 | 106 | |
U15939.1 | - | 5 - 6 | 1387 | 106 | |
U17566.1 | - | 5 - 6 | 1387 | 106 | |
U19720.1 | - | 5 - 6 | 1388 | 106 |