The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114108.1 | NP_001107580.1 | 3 - 4 | 850 | 68 |
NM_017904.3 | NP_060374.2 | 3 - 4 | 850 | 68 | |
XM_011541671.2 | XP_011539973.1 | 3 - 4 | 1961 | 68 | |
XM_017001582.1 | XP_016857071.1 | 3 - 4 | 1984 | 68 | |
GenBank mRNA | AK000626.1 | - | 3 - 4 | 780 | 68 |