The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287135.1 | NP_001274064.1 | 10 - 11 | 1306 | 87 |
NM_001287136.1 | NP_001274065.1 | 9 - 10 | 1059 | 87 | |
NM_001287137.1 | NP_001274066.1 | 8 - 9 | 964 | 87 | |
NM_012395.3 | NP_036527.1 | 9 - 10 | 1130 | 87 | |
XM_005250436.3 | XP_005250493.1 | 11 - 12 | 1105 | 87 | |
XM_005250438.3 | XP_005250495.1 | 9 - 10 | 1052 | 87 | |
XM_005250439.2 | XP_005250496.1 | 10 - 11 | 1863 | 87 | |
XM_011516306.2 | XP_011514608.1 | 10 - 11 | 1674 | 87 | |
XM_017012320.1 | XP_016867809.1 | 11 - 12 | 1238 | 87 | |
XM_017012321.1 | XP_016867810.1 | 9 - 10 | 967 | 87 | |
XM_017012322.1 | XP_016867811.1 | 9 - 10 | 1609 | 87 | |
GenBank mRNA | AB020641.1 | - | 9 - 10 | 1130 | 87 |
AF119833.1 | - | 10 - 11 | 1178 | 87 | |
AK289782.1 | - | 9 - 10 | 988 | 87 | |
AK295086.1 | - | 9 - 10 | 87 | ||
AK297974.1 | - | 10 - 11 | 1262 | 87 | |
AK307484.1 | - | 9 - 10 | 1013 | 87 | |
AK316026.1 | - | 9 - 10 | 1013 | 87 | |
BC136476.1 | - | 9 - 10 | 1200 | 87 | |
BC136477.1 | - | 9 - 10 | 1085 | 87 | |
BC152388.1 | - | 9 - 10 | 989 | 87 | |
BC152436.1 | - | 9 - 10 | 1130 | 87 | |
BC167152.1 | - | 9 - 10 | 1130 | 87 | |
BC167156.1 | - | 9 - 10 | 1130 | 87 |