Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK14
Entrez Gene ID
5218
Gene Name
cyclin dependent kinase 14
Gene Aliases
PFTAIRE1, PFTK1
Chromosome Location
Chr.7: 90595994 - 91210590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287135.1 NP_001274064.1 13 - 14 1557 94
NM_001287136.1 NP_001274065.1 12 - 13 1310 94
NM_001287137.1 NP_001274066.1 11 - 12 1215 94
NM_012395.3 NP_036527.1 12 - 13 1381 94
XM_005250436.3 XP_005250493.1 14 - 15 1356 94
XM_005250438.3 XP_005250495.1 12 - 13 1303 94
XM_005250439.2 XP_005250496.1 13 - 14 2114 94
XM_011516306.2 XP_011514608.1 13 - 14 1925 94
XM_017012320.1 XP_016867809.1 14 - 15 1489 94
XM_017012321.1 XP_016867810.1 12 - 13 1218 94
XM_017012322.1 XP_016867811.1 12 - 13 1860 94
GenBank mRNA AB020641.1 - 12 - 13 1381 94
AF119833.1 - 13 - 14 1429 94
AK289782.1 - 12 - 13 1239 94
AK295086.1 - 12 - 13 1293 94
AK296406.1 - 11 - 12 1169 94
AK297974.1 - 13 - 14 1513 94
AK307484.1 - 12 - 13 1264 94
AK316026.1 - 12 - 13 1264 94
BC136476.1 - 12 - 13 1451 94
BC136477.1 - 12 - 13 1336 94
BC152388.1 - 12 - 13 1240 94
BC152436.1 - 12 - 13 1381 94
BC167152.1 - 12 - 13 1381 94
BC167156.1 - 12 - 13 1381 94

View Full Product Details