The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287135.1 | NP_001274064.1 | 6 - 7 | 904 | 66 |
NM_001287136.1 | NP_001274065.1 | 5 - 6 | 657 | 66 | |
NM_001287137.1 | NP_001274066.1 | 4 - 5 | 562 | 66 | |
NM_012395.3 | NP_036527.1 | 5 - 6 | 728 | 66 | |
XM_005250436.3 | XP_005250493.1 | 7 - 8 | 703 | 66 | |
XM_005250438.3 | XP_005250495.1 | 5 - 6 | 650 | 66 | |
XM_005250439.2 | XP_005250496.1 | 6 - 7 | 1461 | 66 | |
XM_011516306.2 | XP_011514608.1 | 6 - 7 | 1272 | 66 | |
XM_011516307.2 | XP_011514609.1 | 6 - 7 | 1272 | 66 | |
XM_017012320.1 | XP_016867809.1 | 7 - 8 | 836 | 66 | |
XM_017012321.1 | XP_016867810.1 | 5 - 6 | 565 | 66 | |
XM_017012322.1 | XP_016867811.1 | 5 - 6 | 1207 | 66 | |
GenBank mRNA | AB020641.1 | - | 5 - 6 | 728 | 66 |
AF119833.1 | - | 6 - 7 | 776 | 66 | |
AK289782.1 | - | 5 - 6 | 586 | 66 | |
AK295086.1 | - | 5 - 6 | 640 | 66 | |
AK296406.1 | - | 4 - 5 | 516 | 66 | |
AK297974.1 | - | 6 - 7 | 860 | 66 | |
AK307484.1 | - | 5 - 6 | 611 | 66 | |
AK316026.1 | - | 5 - 6 | 611 | 66 | |
BC136476.1 | - | 5 - 6 | 798 | 66 | |
BC136477.1 | - | 5 - 6 | 683 | 66 | |
BC152388.1 | - | 5 - 6 | 587 | 66 | |
BC152436.1 | - | 5 - 6 | 728 | 66 | |
BC167152.1 | - | 5 - 6 | 728 | 66 | |
BC167156.1 | - | 5 - 6 | 728 | 66 |