The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003730.4 | NP_003721.2 | 1 - 2 | 498 | 67 |
XM_017011398.1 | XP_016866887.1 | 1 - 2 | 455 | 67 | |
XM_017011399.1 | XP_016866888.1 | 1 - 2 | 459 | 67 | |
GenBank mRNA | AJ419865.1 | - | 1 - 2 | 452 | 67 |
AJ419866.1 | - | 1 - 2 | 452 | 67 | |
AJ419867.1 | - | 1 - 2 | 452 | 67 | |
AK315467.1 | - | 1 - 2 | 459 | 67 | |
BC001660.2 | - | 1 - 2 | 424 | 67 | |
BC001819.2 | - | 1 - 2 | 424 | 67 | |
BC036282.1 | - | 1 - 2 | 485 | 67 | |
BC039713.1 | - | 1 - 2 | 427 | 67 | |
BC051912.1 | - | 1 - 2 | 437 | 67 | |
U85625.2 | - | 1 - 2 | 455 | 67 |