The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007090.2 | NP_001007091.1 | 1 - 1 | 87 | |
GenBank mRNA | AK058131.1 | - | 1 - 1 | 87 | |
AK058156.1 | - | 1 - 1 | 87 | ||
BC031245.1 | - | 1 - 1 | 87 | ||
CB306127.1 | - | 1 - 1 | 111 | 87 |