The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004499.3 | NP_004490.2 | 5 - 6 | 929 | 97 |
NM_031266.2 | NP_112556.2 | 5 - 6 | 929 | 97 | |
GenBank mRNA | AK054600.1 | - | 4 - 5 | 977 | 97 |
AK097657.1 | - | 5 - 6 | 930 | 97 | |
AK223425.1 | - | 5 - 6 | 906 | 97 | |
BC001616.1 | - | 5 - 6 | 888 | 97 | |
BC002625.2 | - | 5 - 6 | 888 | 97 | |
BC004561.2 | - | 5 - 6 | 896 | 97 | |
BC009359.2 | - | 5 - 6 | 892 | 97 | |
BC036708.2 | - | 5 - 6 | 855 | 97 | |
M65028.1 | - | 5 - 6 | 811 | 97 | |
U76713.1 | - | 4 - 5 | 684 | 97 | |
Z36844.1 | - | 1 - 2 | 97 | 97 |