The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199770.1 | NP_001186699.1 | 1 - 2 | 96 | 76 |
GenBank mRNA | BM720707.1 | - | 1 - 2 | 97 | 76 |