The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024675.3 | NP_078951.2 | 3 - 4 | 407 | 143 |
XM_011545946.2 | XP_011544248.1 | 3 - 4 | 1212 | 143 | |
XM_011545947.2 | XP_011544249.1 | 3 - 4 | 1177 | 143 | |
XM_011545948.2 | XP_011544250.1 | 4 - 5 | 499 | 143 | |
XM_017023671.1 | XP_016879160.1 | 3 - 4 | 1177 | 143 | |
XM_017023672.1 | XP_016879161.1 | 3 - 4 | 407 | 143 | |
XM_017023673.1 | XP_016879162.1 | 3 - 4 | 407 | 143 | |
GenBank mRNA | AK097533.1 | - | 3 - 4 | 356 | 143 |
AK299152.1 | - | 3 - 4 | 304 | 143 | |
BC044254.2 | - | 3 - 4 | 334 | 143 | |
BX647125.1 | - | 3 - 4 | 318 | 143 | |
BX647130.1 | - | 3 - 4 | 360 | 143 | |
CR749637.1 | - | 3 - 4 | 359 | 143 | |
DA192828.1 | - | 3 - 4 | 407 | 143 |