Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH3
Entrez Gene ID
55567
Gene Name
dynein axonemal heavy chain 3
Gene Aliases
DLP3, DNAHC3B, HEL-36, Hsadhc3, dnahc3-b
Chromosome Location
Chr.16: 20933111 - 21159537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017539.2 NP_060009.1 42 - 43 6126 92
XM_006721062.1 XP_006721125.1 42 - 43 6264 92
XM_011545882.2 XP_011544184.1 43 - 44 6704 92
XM_011545883.1 XP_011544185.1 43 - 44 6472 92
XM_011545885.2 XP_011544187.1 43 - 44 6704 92
XM_011545886.2 XP_011544188.1 35 - 36 5079 92
XM_011545888.2 XP_011544190.1 25 - 26 3523 92
XM_011545889.2 XP_011544191.1 25 - 26 3487 92
XM_017023426.1 XP_016878915.1 42 - 43 6569 92
XM_017023427.1 XP_016878916.1 41 - 42 6461 92
XM_017023429.1 XP_016878918.1 43 - 44 6705 92
XM_017023432.1 XP_016878921.1 42 - 43 6264 92
GenBank mRNA AF494040.1 - 42 - 43 6126 92
AK125907.1 - 19 - 20 2843 92

View Full Product Details