The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017539.2 | NP_060009.1 | 47 - 48 | 7023 | 110 |
XM_006721062.1 | XP_006721125.1 | 47 - 48 | 7161 | 110 | |
XM_011545882.2 | XP_011544184.1 | 48 - 49 | 7601 | 110 | |
XM_011545883.1 | XP_011544185.1 | 48 - 49 | 7369 | 110 | |
XM_011545885.2 | XP_011544187.1 | 48 - 49 | 7601 | 110 | |
XM_011545886.2 | XP_011544188.1 | 40 - 41 | 5976 | 110 | |
XM_011545888.2 | XP_011544190.1 | 30 - 31 | 4420 | 110 | |
XM_011545889.2 | XP_011544191.1 | 30 - 31 | 4384 | 110 | |
XM_017023426.1 | XP_016878915.1 | 47 - 48 | 7466 | 110 | |
XM_017023427.1 | XP_016878916.1 | 46 - 47 | 7358 | 110 | |
XM_017023428.1 | XP_016878917.1 | 45 - 46 | 7022 | 110 | |
XM_017023430.1 | XP_016878919.1 | 6 - 7 | 1043 | 110 | |
XM_017023432.1 | XP_016878921.1 | 47 - 48 | 7161 | 110 | |
GenBank mRNA | AF494040.1 | - | 47 - 48 | 7023 | 110 |