Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEC
Entrez Gene ID
5339
Gene Name
plectin
Gene Aliases
EBS1, EBSMD, EBSND, EBSO, EBSOG, EBSPA, HD1, LGMD2Q, PCN, PLEC1, PLEC1b, PLTN
Chromosome Location
Chr.8: 143915147 - 143976800 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000445.4 NP_000436.2 16 - 17 1942 89
NM_201378.3 NP_958780.1 15 - 16 1827 89
NM_201379.2 NP_958781.1 15 - 16 1895 89
NM_201380.3 NP_958782.1 15 - 16 2393 89
NM_201381.2 NP_958783.1 15 - 16 1790 89
NM_201382.3 NP_958784.1 15 - 16 1880 89
NM_201383.2 NP_958785.1 15 - 16 1833 89
NM_201384.2 NP_958786.1 15 - 16 1941 89
XM_005250976.3 XP_005251033.1 16 - 17 2435 89
XM_005250978.3 XP_005251035.1 16 - 17 1960 89
XM_005250979.4 XP_005251036.1 16 - 17 1919 89
XM_005250980.4 XP_005251037.1 16 - 17 2004 89
XM_005250981.3 XP_005251038.1 16 - 17 1841 89
XM_005250982.3 XP_005251039.1 16 - 17 1834 89
XM_005250983.2 XP_005251040.1 15 - 16 1758 89
XM_005250984.4 XP_005251041.1 16 - 17 1823 89
XM_006716588.3 XP_006716651.1 17 - 18 2012 89
XM_006716589.2 XP_006716652.1 16 - 17 1773 89
XM_006716590.3 XP_006716653.1 16 - 17 1883 89
XM_011517130.2 XP_011515432.1 16 - 17 1948 89
XM_011517131.2 XP_011515433.1 15 - 16 1867 89
XM_011517132.2 XP_011515434.1 16 - 17 1956 89
GenBank mRNA AY480044.1 - 15 - 16 1899 89
AY480045.1 - 15 - 16 1770 89
AY480046.1 - 15 - 16 1895 89
AY480047.1 - 15 - 16 2393 89
AY480048.1 - 15 - 16 1790 89
AY480049.1 - 15 - 16 1819 89
AY480050.1 - 15 - 16 1833 89
AY480051.1 - 15 - 16 1941 89
U53204.1 - 16 - 17 1944 89

View Full Product Details