Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHN1
Entrez Gene ID
84069
Gene Name
pleckstrin homology domain containing N1
Gene Aliases
-
Chromosome Location
Chr.1: 965820 - 975108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160184.1 NP_001153656.1 7 - 8 780 93
NM_032129.2 NP_115505.2 7 - 8 744 93
XM_006710944.3 XP_006711007.2 7 - 8 1457 93
XM_011542248.2 XP_011540550.2 6 - 7 1577 93
XM_017002474.1 XP_016857963.1 6 - 7 1577 93
XM_017002475.1 XP_016857964.1 6 - 7 1577 93
XM_017002476.1 XP_016857965.1 6 - 7 1577 93
XM_017002477.1 XP_016857966.1 6 - 7 1577 93
XM_017002478.1 XP_016857967.1 6 - 7 1577 93
GenBank mRNA AL136730.1 - 7 - 8 744 93
BC101386.2 - 7 - 8 775 93
BC101387.2 - 6 - 7 895 93

View Full Product Details