The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303533.1 | NP_001290462.1 | 10 - 11 | 1201 | 76 |
NM_001303534.1 | NP_001290463.1 | 10 - 11 | 1296 | 76 | |
NM_016339.4 | NP_057423.1 | 10 - 11 | 1481 | 76 | |
XM_006721932.3 | XP_006721995.1 | 10 - 11 | 1975 | 76 | |
XM_006721935.3 | XP_006721998.1 | 11 - 12 | 1252 | 76 | |
XM_006721937.3 | XP_006722000.1 | 10 - 11 | 3077 | 76 | |
XM_006721938.3 | XP_006722001.1 | 7 - 8 | 868 | 76 | |
GenBank mRNA | AF117946.1 | - | 10 - 11 | 1116 | 76 |
AK294642.1 | - | 6 - 7 | 1103 | 76 | |
AK304160.1 | - | 10 - 11 | 1296 | 76 | |
AK313492.1 | - | 10 - 11 | 1156 | 76 | |
BC115374.1 | - | 8 - 9 | 1010 | 76 | |
BC115375.1 | - | 8 - 9 | 1010 | 76 |