Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAPGEFL1
Entrez Gene ID
51195
Gene Name
Rap guanine nucleotide exchange factor like 1
Gene Aliases
Link-GEFII
Chromosome Location
Chr.17: 40177010 - 40195656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303533.1 NP_001290462.1 12 - 13 1340 76
NM_001303534.1 NP_001290463.1 12 - 13 1435 76
NM_016339.4 NP_057423.1 12 - 13 1620 76
XM_006721932.3 XP_006721995.1 12 - 13 2114 76
XM_006721935.3 XP_006721998.1 13 - 14 1391 76
XM_006721937.3 XP_006722000.1 12 - 13 3216 76
XM_006721938.3 XP_006722001.1 9 - 10 1007 76
GenBank mRNA AF117946.1 - 12 - 13 1255 76
AK294642.1 - 8 - 9 1242 76
AK304160.1 - 12 - 13 1435 76
AK313492.1 - 12 - 13 1295 76
BC115374.1 - 10 - 11 1149 76
BC115375.1 - 10 - 11 1149 76

View Full Product Details