The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303533.1 | NP_001290462.1 | 9 - 10 | 1104 | 61 |
NM_001303534.1 | NP_001290463.1 | 9 - 10 | 1199 | 61 | |
NM_016339.4 | NP_057423.1 | 9 - 10 | 1384 | 61 | |
XM_006721932.3 | XP_006721995.1 | 9 - 10 | 1878 | 61 | |
XM_006721935.3 | XP_006721998.1 | 10 - 11 | 1155 | 61 | |
XM_006721937.3 | XP_006722000.1 | 9 - 10 | 2980 | 61 | |
XM_006721938.3 | XP_006722001.1 | 6 - 7 | 771 | 61 | |
GenBank mRNA | AF117946.1 | - | 9 - 10 | 1019 | 61 |
AK294642.1 | - | 5 - 6 | 1006 | 61 | |
AK304160.1 | - | 9 - 10 | 1199 | 61 | |
AK313492.1 | - | 9 - 10 | 1059 | 61 | |
BC115374.1 | - | 7 - 8 | 913 | 61 | |
BC115375.1 | - | 7 - 8 | 913 | 61 |