The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256017.2 | NP_001242946.2 | 4 - 5 | 633 | 85 |
GenBank mRNA | AB043820.1 | - | 4 - 5 | 633 | 85 |