The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024728.2 | NP_079004.1 | 5 - 6 | 402 | 143 |
GenBank mRNA | AK021870.1 | - | 5 - 6 | 402 | 143 |