The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011554.2 | NP_001011554.1 | 4 - 5 | 617 | 60 |
NM_001193340.1 | NP_001180269.1 | 4 - 5 | 617 | 60 | |
NM_001193342.1 | NP_001180271.1 | 3 - 4 | 408 | 60 | |
NM_022829.5 | NP_073740.2 | 3 - 4 | 581 | 60 | |
GenBank mRNA | AK056713.1 | - | 3 - 4 | 579 | 60 |
AK096658.1 | - | 4 - 5 | 617 | 60 | |
AK310960.1 | - | 2 - 3 | 591 | 60 | |
AK315131.1 | - | 3 - 4 | 579 | 60 | |
AY072810.3 | - | 3 - 4 | 560 | 60 | |
BC035966.1 | - | 3 - 4 | 579 | 60 | |
CB997336.1 | - | 3 - 4 | 433 | 60 | |
CN310376.1 | - | 4 - 5 | 590 | 60 |