The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006833.4 | NP_006824.2 | 2 - 3 | 229 | 89 |
GenBank mRNA | AK295237.1 | - | 2 - 3 | 207 | 89 |
AK312024.1 | - | 2 - 3 | 220 | 89 | |
BC002520.2 | - | 2 - 3 | 203 | 89 | |
BI113801.1 | - | 2 - 3 | 229 | 89 | |
CB990467.1 | - | 2 - 3 | 253 | 89 | |
U70735.1 | - | 2 - 3 | 151 | 89 |