The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080512.2 | NP_001073981.1 | 12 - 13 | 1721 | 124 |
XM_005270169.4 | XP_005270226.1 | 11 - 12 | 4719 | 124 | |
XM_011540185.2 | XP_011538487.1 | 13 - 14 | 1967 | 124 | |
XM_011540188.2 | XP_011538490.2 | 12 - 13 | 1852 | 124 | |
XM_011540189.2 | XP_011538491.1 | 11 - 12 | 1618 | 124 | |
XM_011540190.2 | XP_011538492.1 | 11 - 12 | 1575 | 124 | |
XM_011540191.2 | XP_011538493.1 | 9 - 10 | 1437 | 124 | |
XM_017016677.1 | XP_016872166.1 | 13 - 14 | 8355 | 124 | |
XM_017016678.1 | XP_016872167.1 | 14 - 15 | 1872 | 124 | |
GenBank mRNA | AK026129.1 | - | 4 - 5 | 623 | 124 |
AK225427.1 | - | 4 - 5 | 623 | 124 | |
AK225429.1 | - | 4 - 5 | 623 | 124 | |
AK307780.1 | - | 6 - 7 | 1006 | 125 | |
AY726586.1 | - | 12 - 13 | 1666 | 124 |