The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014435.1 | NP_001014435.1 | 5 - 6 | 780 | 63 |
NM_005182.2 | NP_005173.1 | 5 - 6 | 628 | 63 | |
XM_005256136.3 | XP_005256193.1 | 5 - 6 | 613 | 63 | |
XM_011523311.1 | XP_011521613.1 | 5 - 6 | 622 | 63 | |
XM_011523312.1 | XP_011521614.1 | 6 - 7 | 701 | 63 | |
GenBank mRNA | AY075019.1 | - | 5 - 6 | 635 | 63 |
AY075020.1 | - | 5 - 6 | 782 | 63 | |
BC033865.1 | - | 5 - 6 | 584 | 63 | |
BI752037.1 | - | 5 - 6 | 762 | 63 |