The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286188.1 | NP_001273117.1 | 4 - 5 | 441 | 92 |
NM_001286189.1 | NP_001273118.1 | 5 - 6 | 405 | 92 | |
NM_001286194.1 | NP_001273123.1 | 4 - 5 | 342 | 92 | |
NM_001286199.1 | NP_001273128.1 | 4 - 5 | 394 | 92 | |
NM_014892.4 | NP_055707.3 | 3 - 4 | 685 | 92 | |
GenBank mRNA | AB029039.1 | - | 3 - 4 | 347 | 92 |
AK295651.1 | - | 4 - 5 | 403 | 92 | |
AK302969.1 | - | 4 - 5 | 327 | 92 | |
AK303001.1 | - | 5 - 6 | 367 | 92 | |
AK307815.1 | - | 5 - 6 | 375 | 92 | |
BC032728.1 | - | 3 - 4 | 647 | 92 | |
BC070071.1 | - | 4 - 5 | 394 | 92 |