The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286188.1 | NP_001273117.1 | 8 - 9 | 1066 | 84 |
NM_001286189.1 | NP_001273118.1 | 9 - 10 | 1030 | 84 | |
NM_001286194.1 | NP_001273123.1 | 8 - 9 | 967 | 84 | |
NM_001286199.1 | NP_001273128.1 | 8 - 9 | 1019 | 84 | |
NM_014892.4 | NP_055707.3 | 7 - 8 | 1310 | 84 | |
GenBank mRNA | AB029039.1 | - | 7 - 8 | 972 | 84 |
AK295651.1 | - | 8 - 9 | 1028 | 84 | |
AK302969.1 | - | 8 - 9 | 952 | 84 | |
AK303001.1 | - | 9 - 10 | 992 | 84 | |
AK307815.1 | - | 9 - 10 | 1000 | 84 | |
BC032728.1 | - | 7 - 8 | 1272 | 84 | |
BC070071.1 | - | 8 - 9 | 1019 | 84 |