The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271867.1 | NP_001258796.1 | 6 - 7 | 526 | 113 |
NM_001271868.1 | NP_001258797.1 | 6 - 7 | 442 | 113 | |
NM_001516.4 | NP_001507.2 | 7 - 8 | 606 | 113 | |
XM_017019228.1 | XP_016874717.1 | 7 - 8 | 524 | 121 | |
GenBank mRNA | AK298128.1 | - | 6 - 7 | 436 | 113 |
AK313200.1 | - | 7 - 8 | 516 | 113 | |
AL558542.3 | - | 7 - 8 | 563 | 113 | |
BC031030.1 | - | 4 - 5 | 267 | 113 | |
BC039726.2 | - | 6 - 7 | 472 | 113 | |
BC047868.2 | - | 6 - 7 | 472 | 113 | |
BC065250.1 | - | 7 - 8 | 499 | 113 | |
DB482432.1 | - | 6 - 7 | 456 | 113 | |
Z30093.1 | - | 6 - 7 | 485 | 113 |