The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271867.1 | NP_001258796.1 | 7 - 8 | 596 | 87 |
NM_001271868.1 | NP_001258797.1 | 7 - 8 | 512 | 87 | |
NM_001516.4 | NP_001507.2 | 8 - 9 | 676 | 87 | |
XM_017019228.1 | XP_016874717.1 | 8 - 9 | 594 | 87 | |
GenBank mRNA | AK298128.1 | - | 7 - 8 | 506 | 87 |
AK313200.1 | - | 8 - 9 | 586 | 87 | |
AL558542.3 | - | 8 - 9 | 87 | ||
BC031030.1 | - | 5 - 6 | 337 | 87 | |
BC039726.2 | - | 7 - 8 | 542 | 87 | |
BC047868.2 | - | 7 - 8 | 542 | 87 | |
BC065250.1 | - | 8 - 9 | 569 | 87 | |
DA953586.1 | - | 7 - 8 | 424 | 87 | |
Z30093.1 | - | 7 - 8 | 555 | 87 |