The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_119376.1 | 41 - 42 | 6301 | 70 | |
GenBank mRNA | AK023521.1 | - | - | 2507 | 70 |
AL833764.1 | - | - | 3589 | 70 | |
AY461812.1 | - | 43 - 44 | 5881 | 70 | |
BX648024.1 | - | 14 - 15 | 3300 | 70 |