Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC6
Entrez Gene ID
319089
Gene Name
tetratricopeptide repeat domain 6
Gene Aliases
C14orf25, NCRNA00291
Chromosome Location
Chr.14: 37595199 - 37842628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001310135.1 NP_001297064.1 30 - 31 6027 136
XM_011537430.2 XP_011535732.1 26 - 27 3859 136
XM_011537431.2 XP_011535733.1 24 - 25 3516 136
XM_017021254.1 XP_016876743.1 29 - 30 5877 136
XM_017021255.1 XP_016876744.1 28 - 29 5742 136
XM_017021256.1 XP_016876745.1 29 - 30 6141 136
GenBank mRNA BC014342.2 - 3 - 4 434 136
BC103914.2 - 11 - 12 1376 136
BC103915.1 - 11 - 12 1376 136
BC103916.2 - 11 - 12 1376 136
BX161415.1 - 10 - 11 1311 136

View Full Product Details