Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC6
Entrez Gene ID
319089
Gene Name
tetratricopeptide repeat domain 6
Gene Aliases
C14orf25, NCRNA00291
Chromosome Location
Chr.14: 37595199 - 37842628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001310135.1 NP_001297064.1 32 - 33 6432 124
XM_011537430.2 XP_011535732.1 28 - 29 4264 124
XM_011537431.2 XP_011535733.1 26 - 27 3921 124
XM_017021254.1 XP_016876743.1 31 - 32 6282 124
XM_017021255.1 XP_016876744.1 30 - 31 6147 124
XM_017021256.1 XP_016876745.1 31 - 32 6546 124
GenBank mRNA BC014342.2 - 5 - 6 839 124
BC103914.2 - 14 - 15 2002 124
BC103915.1 - 13 - 14 1781 124
BC103916.2 - 13 - 14 1781 124
BX161415.1 - 12 - 13 1716 124

View Full Product Details