The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001494.3 | NP_001485.2 | 3 - 4 | 547 | 126 |
XM_017016071.1 | XP_016871560.1 | 3 - 4 | 242 | 126 | |
GenBank mRNA | AB451295.1 | - | 3 - 4 | 256 | 126 |
AB451429.1 | - | 3 - 4 | 256 | 126 | |
AK297177.1 | - | 4 - 5 | 514 | 126 | |
AK309360.1 | - | 4 - 5 | 463 | 126 | |
AK312485.1 | - | 3 - 4 | 432 | 126 | |
BC005145.2 | - | 3 - 4 | 340 | 126 | |
BT006868.1 | - | 3 - 4 | 256 | 126 | |
CR457073.1 | - | 3 - 4 | 256 | 126 | |
D13988.1 | - | 3 - 4 | 276 | 126 | |
EU794614.1 | - | 3 - 4 | 256 | 126 | |
Y13286.1 | - | 3 - 4 | 408 | 126 |