The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001115156.1 | NP_001108628.1 | 4 - 5 | 750 | 137 |
NM_001494.3 | NP_001485.2 | 5 - 6 | 885 | 137 | |
XM_017016071.1 | XP_016871560.1 | 5 - 6 | 580 | 137 | |
GenBank mRNA | AB451295.1 | - | 5 - 6 | 594 | 137 |
AB451429.1 | - | 5 - 6 | 594 | 137 | |
AF144713.1 | - | 2 - 3 | 354 | 137 | |
AK297177.1 | - | 6 - 7 | 852 | 137 | |
AK297554.1 | - | 4 - 5 | 635 | 137 | |
AK309360.1 | - | 6 - 7 | 801 | 137 | |
AK312485.1 | - | 5 - 6 | 770 | 137 | |
BC005145.2 | - | 5 - 6 | 678 | 137 | |
BT006868.1 | - | 5 - 6 | 594 | 137 | |
BX648038.1 | - | 4 - 5 | 431 | 137 | |
CR457073.1 | - | 5 - 6 | 594 | 137 | |
D13988.1 | - | 5 - 6 | 614 | 137 | |
EU794614.1 | - | 5 - 6 | 594 | 137 | |
Y13286.1 | - | 5 - 6 | 746 | 137 |