Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC83
Entrez Gene ID
220047
Gene Name
coiled-coil domain containing 83
Gene Aliases
CT148, HSD9
Chromosome Location
Chr.11: 85855066 - 85920021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286159.1 NP_001273088.1 2 - 3 605 88
NM_173556.4 NP_775827.2 2 - 3 605 88
XM_011544839.2 XP_011543141.1 2 - 3 191 88
XM_011544840.2 XP_011543142.1 2 - 3 263 88
XM_011544841.1 XP_011543143.1 2 - 3 153 88
XM_011544842.1 XP_011543144.1 2 - 3 605 88
XM_011544843.1 XP_011543145.1 2 - 3 605 88
XM_017017354.1 XP_016872843.1 2 - 3 605 88
XM_017017355.1 XP_016872844.1 2 - 3 605 88
GenBank mRNA AK124113.1 - 2 - 3 606 88
AK314036.1 - 2 - 3 309 88
BC040280.1 - 2 - 3 283 88

View Full Product Details