The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006397.2 | NP_006388.2 | 7 - 8 | 67 | |
XM_006722619.2 | XP_006722682.1 | 8 - 9 | 67 | ||
GenBank mRNA | AK315327.1 | - | 7 - 8 | 67 | |
AY363912.1 | - | 7 - 8 | 67 | ||
BC011748.2 | - | 7 - 8 | 67 | ||
BE560339.1 | - | - | 67 | ||
BM664190.1 | - | 1 - 2 | 336 | 67 | |
CB240995.1 | - | 1 - 2 | 336 | 67 | |
Z97029.1 | - | 7 - 8 | 67 |