The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006799.3 | NP_006790.1 | 3 - 4 | 369 | 70 |
NM_144957.2 | NP_659206.1 | 3 - 4 | 369 | 70 | |
XM_017022876.1 | XP_016878365.1 | 3 - 4 | 289 | 70 | |
XM_017022878.1 | XP_016878367.1 | 3 - 4 | 289 | 70 | |
GenBank mRNA | AB031329.1 | - | 3 - 4 | 272 | 70 |
AB031330.1 | - | 3 - 4 | 266 | 70 | |
AB031331.1 | - | 3 - 4 | 352 | 70 | |
AF058300.1 | - | 3 - 4 | 281 | 70 | |
AY359034.1 | - | 3 - 4 | 282 | 70 | |
BC074999.2 | - | 3 - 4 | 305 | 70 | |
BC075000.2 | - | 3 - 4 | 305 | 70 |