Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMRT2
Entrez Gene ID
10655
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.9: 1003237 - 1057554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181872.4 NP_870987.2 3 - 4 828 101
XM_011517687.1 XP_011515989.1 5 - 6 3376 101
XM_011517690.2 XP_011515992.1 4 - 5 1456 101
XM_011517694.2 XP_011515996.1 2 - 3 987 101
XM_017014213.1 XP_016869702.1 5 - 6 1778 101
XM_017014214.1 XP_016869703.1 4 - 5 1453 101
XM_017014215.1 XP_016869704.1 3 - 4 1297 101
GenBank mRNA AF284225.1 - 2 - 3 628 101
BC136493.1 - 2 - 3 707 101
BX647565.1 - 4 - 5 642 101

View Full Product Details