The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300735.1 | NP_001287664.1 | 10 - 11 | 1270 | 116 |
NM_001300736.1 | NP_001287665.1 | 9 - 10 | 1147 | 116 | |
NM_152545.2 | NP_689758.1 | 10 - 11 | 1273 | 116 | |
XM_017007812.1 | XP_016863301.1 | 8 - 9 | 991 | 116 | |
XM_017007813.1 | XP_016863302.1 | 10 - 11 | 1361 | 116 | |
GenBank mRNA | AK056257.1 | - | 10 - 11 | 1252 | 116 |
AK291459.1 | - | 10 - 11 | 1218 | 116 | |
AK303625.1 | - | 9 - 10 | 1122 | 116 | |
BC121003.1 | - | 10 - 11 | 1185 | 116 | |
BC121004.1 | - | 9 - 10 | 1062 | 116 |