The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145294.4 | NP_660337.3 | 37 - 38 | 4797 | 78 |
XM_017023023.1 | XP_016878512.1 | 38 - 39 | 5067 | 78 | |
XM_017023024.1 | XP_016878513.1 | 20 - 21 | 2680 | 78 | |
GenBank mRNA | AK092719.1 | - | 8 - 9 | 1513 | 78 |
AK093609.1 | - | 3 - 4 | 1123 | 78 | |
AK131510.1 | - | 20 - 21 | 3103 | 78 |