Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
Chromosome Location
Chr.11: 78100942 - 78139660 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007027.2 NP_001007028.1 11 - 12 1339 225
NM_024079.4 NP_076984.2 11 - 12 1339 150
XM_005274247.3 XP_005274304.1 11 - 12 2100 150
XM_011545252.2 XP_011543554.1 11 - 12 1349 150
XM_017018274.1 XP_016873763.1 11 - 12 2087 225
GenBank mRNA AJ224875.1 - 11 - 12 1293 150
AK075184.1 - 11 - 12 1279 225
AK075194.1 - 11 - 12 1294 150
AK098614.1 - 11 - 12 1339 150
BC001133.2 - 11 - 12 1322 150
DB362104.1 - 2 - 3 150
KU178764.1 - 11 - 12 1274 150
KU178765.1 - 3 - 4 331 150

View Full Product Details