The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007027.2 | NP_001007028.1 | 5 - 6 | 608 | 136 |
NM_024079.4 | NP_076984.2 | 5 - 6 | 608 | 136 | |
XM_005274247.3 | XP_005274304.1 | 5 - 6 | 1369 | 136 | |
XM_011545252.2 | XP_011543554.1 | 5 - 6 | 618 | 136 | |
XM_017018274.1 | XP_016873763.1 | 5 - 6 | 1356 | 136 | |
GenBank mRNA | AJ224875.1 | - | 5 - 6 | 562 | 136 |
AK075184.1 | - | 5 - 6 | 548 | 136 | |
AK098614.1 | - | 5 - 6 | 608 | 136 | |
BC001133.2 | - | 5 - 6 | 591 | 136 | |
KU178764.1 | - | 5 - 6 | 543 | 136 |